missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-4-methoxy-3(2H)-furanone 97.0+%, TCI America™

Catalog No. D34995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34995G 5 g
D349925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34995G Supplier TCI America Supplier No. D34995G
Only null left

Chemical Identifiers

CAS 4077-47-8
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00209504
InChI Key SIMKGHMLPVDSJE-UHFFFAOYNA-N
PubChem CID 61325
IUPAC Name 4-methoxy-2,5-dimethyl-2,3-dihydrofuran-3-one
SMILES COC1=C(C)OC(C)C1=O

Specifications

Color Yellow
Boiling Point 63°C
Quantity 5 g
Formula Weight 142.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-4-methoxy-3(2H)-furanone