accessibility menu, dialog, popup

UserName

FNBr

CAS RN 407-20-5

IUPAC Name: 3-bromo-5-fluoropyridine
Synonyms: 3-bromo-5-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: HNNNBQRRIHKFLI-UHFFFAOYSA-N
SMILES: FC1=CN=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)