accessibility menu, dialog, popup

UserName

CAS RN 4068-75-1

IUPAC Name: methyl 2-hydroxy-5-iodobenzoate
Synonyms: methyl 2-hydroxy-5-iodobenzoate
Molecular Weight (g/mol): 278.05
Molecular Formula: C8H7IO3
InChi Key: NRSWJTRJHPRZMH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(I)=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)