accessibility menu, dialog, popup

UserName

CAS RN 4050-45-7

IUPAC Name: (2E)-hex-2-ene
Synonyms: 2-hexene (trans) 4-octene (trans) hexene - mixture of isomers
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: RYPKRALMXUUNKS-HWKANZROSA-N
SMILES: CCC\C=C\C
Boiling Point 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)