missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexene, 99%

Catalog No. AAB2228988
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2228988 2.5 g
AAB2228909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2228988 Supplier Thermo Scientific Chemicals Supplier No. B2228988
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4050-45-7 | C6H12 | 84.16 g/mol

trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-2-Hexene is used as a co monomer in the production of polyethylene. The use of trans-2-hexene, which is derived from 3-hexyne using sodium in liquid ammonia.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4050-45-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
PubChem CID 639661
SMILES CCC\C=C\C

Specifications

Melting Point -99°C
Density 0.674
Boiling Point 68°C to 69°C
Flash Point −20°C (−4°F)
Odor Mild
Refractive Index 1.394
Quantity 2.5 g
UN Number UN3295
Beilstein 1719020
Solubility Information Difficult to mix.
IUPAC Name (2E)-hex-2-ene
Formula Weight 84.16
Percent Purity 99%
Chemical Name or Material trans-2-Hexene
Show More Show Less
Hazard Category H225-H304-H315-H319
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 223-752-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only