missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexene 99.0+%, TCI America™

Catalog No. H03805ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03805ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H03805ML Supplier TCI America Supplier No. H03805ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4050-45-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00009473
InChI Key RYPKRALMXUUNKS-HWKANZROSA-N
Synonym trans-2-hexene, e-2-hexene, 2-hexene, e, hexene, isomer, hexene, isomers, unii-2tz30ggg1a, 2e-hexene, e-hex-2-ene, 2tz30ggg1a, 2-hexene, 2e
PubChem CID 639661
IUPAC Name (2E)-hex-2-ene
SMILES CCC\C=C\C

Specifications

Color Yellow
Boiling Point 68°C
Quantity 5 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-2-Hexene
EINECSNumber (2)-0031
TSCA No