accessibility menu, dialog, popup

UserName

CH3HNNHOHNOH(E/Z)

CAS RN 404950-80-7

IUPAC Name: N-hydroxy-3-[4-({[2-(2-methyl-1H-indol-3-yl)ethyl]amino}methyl)phenyl]prop-2-enamide
Synonyms: panobinostat
Molecular Weight (g/mol): 349.43
Molecular Formula: C21H23N3O2
InChi Key: FPOHNWQLNRZRFC-UHFFFAOYSA-N
SMILES: CC1=C(CCNCC2=CC=C(C=CC(=O)NO)C=C2)C2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)