accessibility menu, dialog, popup

UserName

NNNH2

CAS RN 40230-24-8

IUPAC Name: 4,6-diphenylpyrimidin-2-amine
Synonyms: 4,6-diphenylpyrimidin-2-amine
Molecular Weight (g/mol): 247.3
Molecular Formula: C16H13N3
InChi Key: KZUCBEYDRUCBCS-UHFFFAOYSA-N
SMILES: NC1=NC(=CC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results