missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-diphenylpyrimidine 98.0+%, TCI America™

Catalog No. A29215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29215G 5 g
A29211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29215G Supplier TCI America Supplier No. A29215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40230-24-8
Molecular Formula C16H13N3
Molecular Weight (g/mol) 247.301
MDL Number MFCD00234910
InChI Key KZUCBEYDRUCBCS-UHFFFAOYSA-N
PubChem CID 619024
IUPAC Name 4,6-diphenylpyrimidin-2-amine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)N)C3=CC=CC=C3

Specifications

Melting Point 136°C
Quantity 5 g
Formula Weight 247.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-diphenylpyrimidine