accessibility menu, dialog, popup

UserName

CAS RN 4023-34-1

IUPAC Name: cyclopropanecarbonyl chloride
Synonyms: cyclopropanecarbonyl chloride
Molecular Weight (g/mol): 104.53
Molecular Formula: C4H5ClO
InChi Key: ZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILES: ClC(=O)C1CC1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)