missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonyl Chloride 97.0+%, TCI America™

Catalog No. C1059500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1059500G 500 g
C105925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1059500G Supplier TCI America Supplier No. C1059500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4023-34-1
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00001277
InChI Key ZOOSILUVXHVRJE-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
PubChem CID 77637
IUPAC Name cyclopropanecarbonyl chloride
SMILES C1CC1C(=O)Cl

Specifications

Boiling Point 119°C
Quantity 500 g
UN Number 2920
Formula Weight 104.53
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Cyclopropanecarbonyl Chloride
TSCA Yes
Recommended Storage Refrigerator