accessibility menu, dialog, popup

UserName

CH3NNH3CH3COSOOO

CAS RN 401788-98-5

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate
Synonyms: 1-butyl-3-methylimidazolium methyl sulfate(1-)
Molecular Weight (g/mol): 250.31
Molecular Formula: C9H18N2O4S
InChi Key: MEMNKNZDROKJHP-UHFFFAOYSA-M
SMILES: COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)