missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Methyl Sulfate 98.0+%, TCI America™

Catalog No. B55275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55275G 5 g
B552725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55275G Supplier TCI America Supplier No. B55275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 401788-98-5
Molecular Formula C9H18N2O4S
Molecular Weight (g/mol) 250.31
MDL Number MFCD03095437
InChI Key MEMNKNZDROKJHP-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate
PubChem CID 12095228
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate
SMILES COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1

Specifications

Melting Point -4°C
Quantity 5 g
UN Number 3265
Formula Weight 250.31
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Methyl Sulfate