missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium methyl sulfate, 99%

Catalog No. AAH2775418
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2775418 50 g
AAH2775406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2775418 Supplier Thermo Scientific Chemicals Supplier No. H2775418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 401788-98-5 | C9H18N2O4S | 250.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401788-98-5
Molecular Formula C9H18N2O4S
Molecular Weight (g/mol) 250.31
MDL Number MFCD03095437
InChI Key MEMNKNZDROKJHP-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate
PubChem CID 12095228
SMILES COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1

Specifications

Density 1.21
Refractive Index 1.478
Quantity 50 g
UN Number UN3265
IUPAC Name 1-butyl-3-methylimidazol-3-ium;methyl sulfate
Formula Weight 250.32
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium methyl sulfate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only