accessibility menu, dialog, popup

UserName

CAS RN 398-23-2

IUPAC Name: 4,4'-difluoro-1,1'-biphenyl
Synonyms: 4,4'-difluorobiphenyl
Molecular Weight (g/mol): 190.19
Molecular Formula: C12H8F2
InChi Key: PZDAAZQDQJGXSW-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=C(F)C=C1
Boiling Point 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)