accessibility menu, dialog, popup

UserName

CAS RN 3967-54-2

ClOOO

CAS RN 3967-54-2

IUPAC Name: 4-chloro-1,3-dioxolan-2-one
Synonyms: 4-chloro-1,3-dioxolan-2-one
Molecular Weight (g/mol): 122.5
Molecular Formula: C3H3ClO3
InChi Key: OYOKPDLAMOMTEE-UHFFFAOYNA-N
SMILES: ClC1COC(=O)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results