missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,3-dioxolan-2-one 95.0+%, TCI America™

Catalog No. p-7125189
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18585G 5 g
C185825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18585G Supplier TCI America Supplier No. C18585G
Only null left

Chemical Identifiers

CAS 3967-54-2
Molecular Formula C3H3ClO3
Molecular Weight (g/mol) 122.504
MDL Number MFCD00005383
InChI Key OYOKPDLAMOMTEE-UHFFFAOYSA-N
Synonym 4-Chloro-2-oxo-1,3-dioxolane, Chloroethylene Carbonate
PubChem CID 98075
IUPAC Name 4-chloro-1,3-dioxolan-2-one
SMILES C1C(OC(=O)O1)Cl

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 122.50
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-1,3-dioxolan-2-one
EINECSNumber (5)-6663
TSCA No
Recommended Storage Refrigerator