accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 3938-96-3

IUPAC Name: ethyl 2-methoxyacetate
Synonyms: ethyl 2-methoxyacetate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: JLEKJZUYWFJPMB-UHFFFAOYSA-N
SMILES: CCOC(=O)COC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)