missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methoxyacetate, 98%

Catalog No. AAA1877018
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1877018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1877018 Supplier Thermo Scientific Chemicals Supplier No. A1877018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3938-96-3 | C5H10O3 | 118.132 g/mol

Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 3938-96-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00009176
InChI Key JLEKJZUYWFJPMB-UHFFFAOYSA-N
Synonym ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
PubChem CID 77544
IUPAC Name ethyl 2-methoxyacetate
SMILES CCOC(=O)COC

Specifications

Density 1
Boiling Point 44°C to 46°C (9 mmHg)
Flash Point 46°C (115°F)
Refractive Index 1.401
Quantity 50 g
UN Number UN3272
Beilstein 1744761
Solubility Information Slightly soluble in water.
Formula Weight 118.13
Percent Purity 98%
Chemical Name or Material Ethyl methoxyacetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 223-521-7
TSCA No

RUO – Research Use Only