missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methoxyacetate 98.0+%, TCI America™

Catalog No. E09795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09795G 5 g
E097925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09795G Supplier TCI America Supplier No. E09795G

Chemical Identifiers

CAS 3938-96-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00009176
InChI Key JLEKJZUYWFJPMB-UHFFFAOYSA-N
Synonym ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
PubChem CID 77544
IUPAC Name ethyl 2-methoxyacetate
SMILES CCOC(=O)COC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 g
UN Number 3272
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Methoxyacetate