accessibility menu, dialog, popup

UserName

NH2FFFF

CAS RN 393-39-5

IUPAC Name: 4-fluoro-2-(trifluoromethyl)aniline
Synonyms: 4-fluoro-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: LRCQLCWUUBSUOY-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results