accessibility menu, dialog, popup

UserName

CAS RN 39215-21-9

IUPAC Name: [1,1'-binaphthalene]-2,2',3,3'-tetrol
Synonyms: [1,1'-binaphthalene]-2,2',3,3'-tetrol
Molecular Weight (g/mol): 318.33
Molecular Formula: C20H14O4
InChi Key: OOSHJGKXQBJASF-UHFFFAOYSA-N
SMILES: OC1=C(O)C(=C2C=CC=CC2=C1)C1=C2C=CC=CC2=CC(O)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results