accessibility menu, dialog, popup

UserName

CAS RN 39150-45-3

NH2BrBrSNH2OO

CAS RN 39150-45-3

IUPAC Name: 4-amino-3,5-dibromobenzene-1-sulfonamide
Synonyms: 4-amino-3,5-dibromobenzenesulfonamide
Molecular Weight (g/mol): 329.99
Molecular Formula: C6H6Br2N2O2S
InChi Key: DLXJPWSYRLLYSV-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1Br)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results