missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosulfanilamide 97.0+%, TCI America™

Catalog No. D021225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D021225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D021225G Supplier TCI America Supplier No. D021225G
Only null left

Chemical Identifiers

CAS 39150-45-3
Molecular Formula C6H6Br2N2O2S
Molecular Weight (g/mol) 329.994
MDL Number MFCD00014783
InChI Key DLXJPWSYRLLYSV-UHFFFAOYSA-N
PubChem CID 3084733
IUPAC Name 4-amino-3,5-dibromobenzenesulfonamide
SMILES C1=C(C=C(C(=C1Br)N)Br)S(=O)(=O)N

Specifications

Melting Point 242°C
Quantity 25 g
Formula Weight 329.99
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromosulfanilamide