accessibility menu, dialog, popup

UserName

CAS RN 3913-81-3

IUPAC Name: (2E)-dec-2-enal
Synonyms: trans-2-decenal
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: MMFCJPPRCYDLLZ-CMDGGOBGSA-N
SMILES: CCCCCCC\C=C\C=O
Percent Purity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)