missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Decenal, ≥96% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018535
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018535 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018535 Supplier Merck Emd Millipore Supplier No. 913091ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 3913-81-3
Density 0.841 g/mL (at 25°C)
Linear Formula CH3(CH2)6 CH=CHCHO
MDL Number MFCD00014679
Boiling Point 78°C to 80°C (3 mmHg)
Type Analytical Standard
Quantity 1 mL
Formula Weight 154.25
Percent Purity ≥96% (GC)
Grade Analytical Standard