accessibility menu, dialog, popup

UserName

H3CH3CCH3NNClNOHH3CH3CCH3

CAS RN 3864-99-1

IUPAC Name: 2,4-di-tert-butyl-6-(5-chloro-2H-1,2,3-benzotriazol-2-yl)phenol
Synonyms: 2,4-di-tert-butyl-6-(5-chloro-1,2,3-benzotriazol-2-yl)phenol
Molecular Weight (g/mol): 357.88
Molecular Formula: C20H24ClN3O
InChi Key: UWSMKYBKUPAEJQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N2N=C3C=CC(Cl)=CC3=N2)=C(O)C(=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results