missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Carboxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. C10235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10235G Supplier TCI America Supplier No. C10235G

Chemical Identifiers

CAS 38628-51-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00016560
InChI Key QSPCOYVUYYFWAU-UHFFFAOYSA-N
Synonym 4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
PubChem CID 854054
IUPAC Name 4-(2-carboxyethyl)benzoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Carboxyphenyl)propionic Acid