missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Carboxyphenyl)propionic acid, 98%

Catalog No. AAA1508122
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1508122 100 g
AAA1508106 5 g
AAA1508109 10 g
AAA1508114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1508122 Supplier Thermo Scientific Chemicals Supplier No. A1508122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38628-51-2 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38628-51-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00016560
InChI Key QSPCOYVUYYFWAU-UHFFFAOYSA-N
Synonym 4-2-carboxyethyl benzoic acid, 3-4-carboxyphenyl propionic acid, 3-4-carboxyphenyl propanoic acid, benzenepropanoic acid, 4-carboxy, pubchem19268, acmc-1ctf7, 4-carboxyhydrocinnamic acid, 4-carboxybenzenepropionic acid, 4-2-carboxyethyl benzoicacid, carboxyphenylpropionicacid; 985
PubChem CID 854054
IUPAC Name 4-(2-carboxyethyl)benzoic acid
SMILES OC(=O)CCC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 288°C to 291°C
Quantity 100 g
Beilstein 3271796
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material 3-(4-Carboxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only