CAS RN 38609-97-1
CAS RN 38609-97-1
IUPAC Name:
sodium 2-(9-oxo-9,10-dihydroacridin-10-yl)acetate
Synonyms:
sodium 2-(9-oxoacridin-10-yl)acetate
Molecular Weight (g/mol):
275.24
Molecular Formula:
C15H10NNaO3
InChi Key:
FQMLTEAEJZVTAJ-UHFFFAOYSA-M
SMILES:
[Na+].[O-]C(=O)CN1C2=CC=CC=C2C(=O)C2=CC=CC=C12
1
–
1
of
1
results
1
9-Oxoacridine-10-acetic Acid 98.0+%, TCI America™
CAS: 38609-97-1 Molecular Formula: C15H11NO3 Molecular Weight (g/mol): 253.257 MDL Number: MFCD00157050 InChI Key: UOMKBIIXHQIERR-UHFFFAOYSA-N Synonym: 10-(Carboxymethyl)-9(10H)-acridone, 9-Oxo-10(9H)-acridineacetic Acid, Cridanimod PubChem CID: 38072 IUPAC Name: 2-(9-oxoacridin-10-yl)acetic acid SMILES: C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2CC(=O)O