accessibility menu, dialog, popup

UserName

CAS RN 38465-60-0

IUPAC Name: copper(2+) bis(tetrafluoroboranuide)
Synonyms: copper(II) tetrafluoroborate
Molecular Weight (g/mol): 237.15
Molecular Formula: B2CuF8
InChi Key: HMUNZEYTSRPVBE-UHFFFAOYSA-N
SMILES: [Cu++].F[B-](F)(F)F.F[B-](F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results