missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Tetrafluoroborate (ca. 45% in Water), TCI America™

Catalog No. C2410500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2410500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2410500G Supplier TCI America Supplier No. C2410500G

Chemical Identifiers

CAS 38465-60-0
Molecular Formula B2CuF8
Molecular Weight (g/mol) 237.15
MDL Number MFCD00016054
InChI Key HMUNZEYTSRPVBE-UHFFFAOYSA-N
Synonym copper fluoroborate, copper fluoborate, copper ii tetrafluoroborate, copper ditetrafluoroborate, cupric tetrafluoroborate hydrate, copper ii tetrafluoroborate in h2o, cupric fluoborate, copper ii fluoborate, copper 2+ ditetrafluoroborate, copper 2+ tetrafluoroborate 1-
PubChem CID 170058
IUPAC Name copper(2+) bis(tetrafluoroboranuide)
SMILES [Cu++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue
Quantity 500 g
UN Number 3264
Formula Weight 237.15
Physical Form Liquid
Chemical Name or Material Copper(II) Tetrafluoroborate (ca. 45% in Water)
EINECSNumber (1)-0043
TSCA Yes