missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) tetrafluoroborate, 45% aq. soln.

Catalog No. AA89666A1
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA89666A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA89666A1 Supplier Thermo Scientific Chemicals Supplier No. 089666A1

CAS: 38465-60-0 | B2CuF8 | 237.15 g/mol

Copper(II) tetrafluoroborate is used in copper plating and in the production of printed circuits. It serves as an intermediate for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. It is also employed in the promotion of Diels-Alder reactions and decomposition of diazo compounds. Further, it is used as a component of fluxing and plating, as a catalyst and as an active filler in resin bonded abrasives. It is used in flame-retardant production, metal treatment, grain refining agents, electrolytic generation of boron and in the preparation of glazing frits. In addition it functions as an oxidizing agent and reacts with siloxycyclopropanes to generate beta acylalkylcopper(II) species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38465-60-0
Molecular Formula B2CuF8
Molecular Weight (g/mol) 237.15
MDL Number MFCD00016054
InChI Key HMUNZEYTSRPVBE-UHFFFAOYSA-N
Synonym copper fluoroborate, copper fluoborate, copper ii tetrafluoroborate, copper ditetrafluoroborate, cupric tetrafluoroborate hydrate, copper ii tetrafluoroborate in h2o, cupric fluoborate, copper ii fluoborate, copper 2+ ditetrafluoroborate, copper 2+ tetrafluoroborate 1-
PubChem CID 170058
IUPAC Name copper;ditetrafluoroborate
SMILES [Cu++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue
Physical Form Liquid
Quantity 1 kg
UN Number UN3264
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 237.15
Concentration or Composition (by Analyte or Components) 45% aq. soln.
Chemical Name or Material Copper (II) tetrafluoroborate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 253-959-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only