accessibility menu, dialog, popup

UserName

CAS RN 38402-02-7

ClCH3NCH3OHHO

CAS RN 38402-02-7

IUPAC Name: bis(2-hydroxyethyl)dimethylazanium chloride
Synonyms: bis(2-hydroxyethyl)dimethylazanium chloride
Molecular Weight (g/mol): 169.65
Molecular Formula: C6H16ClNO2
InChi Key: OPVXPOIGBSHXNM-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(CCO)CCO
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)