missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl)dimethylammonium Chloride 98.0+%, TCI America™

Catalog No. B43845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43845G 5 g
B438425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43845G Supplier TCI America Supplier No. B43845G

Chemical Identifiers

CAS 38402-02-7
Molecular Formula C6H16ClNO2
Molecular Weight (g/mol) 169.649
MDL Number MFCD00041987
InChI Key OPVXPOIGBSHXNM-UHFFFAOYSA-M
Synonym bis 2-hydroxyethyl dimethylammonium chloride, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethylethanaminium chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride 1:1, bis 2-hydroxyethyl dimethylazanium chloride, bis 2-hydroxyethyl-dimethylazanium chloride, n-2-hydroxyethyl-n,n-dimethyl-n-2-hydroxyethyl ammonium chloride
PubChem CID 162303
IUPAC Name bis(2-hydroxyethyl)-dimethylazanium;chloride
SMILES C[N+](C)(CCO)CCO.[Cl-]

Specifications

Color White
Quantity 5 g
Formula Weight 169.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-hydroxyethyl)dimethylammonium Chloride