missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl)dimethylammonium chloride, 99%

Catalog No. AC402530250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402530250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402530250 Supplier Thermo Scientific Chemicals Supplier No. 402530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 38402-02-7 | C6H16ClNO2 | 169.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 38402-02-7
Molecular Formula C6H16ClNO2
Molecular Weight (g/mol) 169.65
MDL Number MFCD00041987
InChI Key OPVXPOIGBSHXNM-UHFFFAOYSA-M
Synonym bis 2-hydroxyethyl dimethylammonium chloride, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethylethanaminium chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride 1:1, bis 2-hydroxyethyl dimethylazanium chloride, bis 2-hydroxyethyl-dimethylazanium chloride, n-2-hydroxyethyl-n,n-dimethyl-n-2-hydroxyethyl ammonium chloride
PubChem CID 162303
IUPAC Name bis(2-hydroxyethyl)-dimethylazanium;chloride
SMILES C[N+](C)(CCO)CCO.[Cl-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2N(Cl)(CH3)2
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 169.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-913-3

RUO – Research Use Only