accessibility menu, dialog, popup

UserName

H3COOFFCl

CAS RN 383-62-0

IUPAC Name: ethyl 2-chloro-2,2-difluoroacetate
Synonyms: ethyl 2-chloro-2,2-difluoroacetate
Molecular Weight (g/mol): 158.53
Molecular Formula: C4H5ClF2O2
InChi Key: GVCAWQUJCHZRCB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(F)(F)Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)