missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chlorodifluoroacetate, 98%

Catalog No. AAA1423222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423222 100 g
AAA1423214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1423222 Supplier Thermo Scientific Chemicals Supplier No. A1423222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 383-62-0 | C4H5ClF2O2 | 158.529 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 383-62-0
Molecular Formula C4H5ClF2O2
Molecular Weight (g/mol) 158.529
MDL Number MFCD00013662
InChI Key GVCAWQUJCHZRCB-UHFFFAOYSA-N
Synonym ethyl chlorodifluoroacetate, chlorodifluoroacetic acid ethyl ester, ethyl chlorodifluoroethanoate, acetic acid, chlorodifluoro-, ethyl ester, ethylchlorodifluoroacetate, acetic acid, 2-chloro-2,2-difluoro-, ethyl ester, ethyl 2-chloro-2,2-difluoro-acetate, pubchem12573, ethyl chlordifluoroacetate, acmc-1cr8n
PubChem CID 67843
IUPAC Name ethyl 2-chloro-2,2-difluoroacetate
SMILES CCOC(=O)C(F)(F)Cl

Specifications

Density 1.252
Boiling Point 96°C to 97°C
Flash Point 18°C (64°F)
Refractive Index 1.358
Quantity 100 g
UN Number UN2924
Beilstein 1761413
Sensitivity Moisture sensitive
Formula Weight 158.53
Percent Purity 98%
Chemical Name or Material Ethyl chlorodifluoroacetate
Show More Show Less
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-850-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only