accessibility menu, dialog, popup

UserName

CAS RN 3825-26-1

NH3HOOFFFFFFFFFFFFFFF

CAS RN 3825-26-1

IUPAC Name: pentadecafluorooctanoic acid amine
Synonyms: perfluorooctanoic acid amine
Molecular Weight (g/mol): 431.1
Molecular Formula: C8H4F15NO2
InChi Key: YOALFLHFSFEMLP-UHFFFAOYSA-N
SMILES: N.OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results