CAS RN 380427-38-3
IUPAC Name:
[4-(propan-2-ylsulfanyl)phenyl]boronic acid
Synonyms:
4-(isopropylsulfanyl)phenylboronic acid
Molecular Weight (g/mol):
196.07
Molecular Formula:
C9H13BO2S
InChi Key:
FYJDSWBEPMIWEC-UHFFFAOYSA-N
SMILES:
CC(C)SC1=CC=C(C=C1)B(O)O