missing translation for 'onlineSavingsMsg'
Learn More

4-(Isopropylthio)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. I09501G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. I09501G missing translation for 'mfr' TCI America missing translation for 'supplierNo' I09501G

Specifications

Chemical Name or Material 4-(Isopropylthio)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 380427-38-3
Melting Point 88°C
Color White
Molecular Formula C9H13BO2S
Quantity 1 g
MDL Number MFCD03411947
Synonym 4-isopropylthiophenylboronic acid, 4-isopropylthio phenylboronic acid, 4-isopropylthio benzeneboronic acid, 4-isopropylsulfanyl phenylboronic acid, boronic acid, 4-1-methylethyl thio phenyl, 4-iso-propylthiobenzeneboronic acid, 4-isopropylthio phenyl boronic acid, 4-isopropylsulfanyl phenyl boronic acid, 4-propan-2-ylsulfanyl phenyl boronic acid
InChI Key FYJDSWBEPMIWEC-UHFFFAOYSA-N
SMILES CC(C)SC1=CC=C(C=C1)B(O)O
IUPAC Name [4-(propan-2-ylsulfanyl)phenyl]boronic acid
Molecular Weight (g/mol) 196.07
PubChem CID 2773483
Formula Weight 196.07
Physical Form Crystalline Powder
Show More Show Less