accessibility menu, dialog, popup

UserName

CAS RN 3800-06-4

OFH2N

CAS RN 3800-06-4

IUPAC Name: 2-(4-fluorobenzoyl)aniline
Synonyms: 2-(4-fluorobenzoyl)aniline
Molecular Weight (g/mol): 215.23
Molecular Formula: C13H10FNO
InChi Key: FFFXIQFESQNINT-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results