missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10611G Supplier TCI America Supplier No. F10611G

Specifications

Chemical Name or Material 4-Fluoro-3-formylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 374538-01-9
Melting Point 220°C
Color White-Yellow
Molecular Formula C7H6BFO3
Quantity 1 g
MDL Number MFCD02093074
Synonym 4-fluoro-3-formylbenzeneboronic acid, 4-fluoro-3-formylphenyl boronic acid, boronic acid, 4-fluoro-3-formylphenyl, 4-fluoro-3-formyl-phenyl boronic acid, pubchem10809, acmc-1cswy, 3-formyl 4-fluorophenyl boronic acid, 5-borono-2-fluorobenzaldehyde, 3-formyl-4-fluorophenyl boronic acid, 4-fluoro-3-formyl benzene boronic acid
InChI Key YABSTJQEBSKPCG-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)F)C=O)(O)O
IUPAC Name (4-fluoro-3-formylphenyl)boronic acid
Molecular Weight (g/mol) 167.93
PubChem CID 2778651
Formula Weight 167.93
Physical Form Crystalline Powder
Show More Show Less