accessibility menu, dialog, popup

UserName

CAS RN 373384-14-6

BONCH3H3CHOOH

CAS RN 373384-14-6

IUPAC Name: [3-(dimethylcarbamoyl)phenyl]boronic acid
Synonyms: 3-(dimethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: DCXXIDMHTQDSLY-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CC(=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results