missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D41941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41941G Supplier TCI America Supplier No. D41941G

Specifications

Chemical Name or Material 3-(Dimethylcarbamoyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 373384-14-6
Color White
Molecular Formula C9H12BNO3
Quantity 1 g
MDL Number MFCD03412066
Synonym 3-dimethylcarbamoyl phenyl boronic acid, 3-dimethylcarbamoyl phenylboronic acid, 3-n,n-dimethylaminocarbonyl phenylboronic acid, n,n-dimethylbenzamide-3-boronic acid, 3-n,n-dimethylaminocarbonyl benzene boronic acid, 3-dimethylaminocarbonyl benzeneboronic acid, 3-dimethylcarbamoyl benzeneboronic acid, n,n-dimethyl 3-boronobenzamide, boronic acid, 3-dimethylamino carbonyl phenyl
InChI Key DCXXIDMHTQDSLY-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C(=O)N(C)C)(O)O
IUPAC Name [3-(dimethylcarbamoyl)phenyl]boronic acid
Molecular Weight (g/mol) 193.009
PubChem CID 2773392
Formula Weight 193.01
Physical Form Crystalline Powder
Show More Show Less