CAS RN 372956-40-6
IUPAC Name:
3,5-bis({4'-fluoro-[1,1'-biphenyl]-4-yl})-4'-(4-fluorophenyl)-1,1'-biphenyl
Synonyms:
3,5-bis({4'-fluoro-[1,1'-biphenyl]-4-yl})-4'-(4-fluorophenyl)-1,1'-biphenyl
Molecular Weight (g/mol):
588.67
Molecular Formula:
C42H27F3
InChi Key:
KMIGKTQTBLIAJV-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)C1=CC=C(F)C=C1)C1=CC=C(C=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
1,3,5-Tris(4'-fluorobiphenyl-4-yl)benzene 95.0+%, TCI America™
CAS: 372956-40-6 Molecular Formula: C42H27F3 Molecular Weight (g/mol): 588.673 InChI Key: KMIGKTQTBLIAJV-UHFFFAOYSA-N Synonym: 4,4′′′′-Difluoro-5′′-(4′-fluoro-[1,1′-biphenyl]-4-yl)-1,1′:4′,1′′:3′′,1′′′:4′′′,1′′′′-quinquephenyl PubChem CID: 11467407 IUPAC Name: 1,3,5-tris[4-(4-fluorophenyl)phenyl]benzene SMILES: C1=CC(=CC=C1C2=CC=C(C=C2)F)C3=CC(=CC(=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)F)C6=CC=C(C=C6)C7=CC=C(C=C7)F