accessibility menu, dialog, popup

UserName

CAS RN 37149-18-1

CH3H3CH3CH3CNOOHO

CAS RN 37149-18-1

IUPAC Name: (4-carboxy-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: 4-carboxy-2,2,6,6-tetramethylpiperidin-1-yloxidanyl
Molecular Weight (g/mol): 200.26
Molecular Formula: C10H18NO3
InChi Key: CYQGCJQJIOARKD-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results