missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 97.0+%, TCI America™

Catalog No. p-7124973
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1428100MG 100 mg
C14281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1428100MG Supplier TCI America Supplier No. C1428100MG
Only null left

Chemical Identifiers

CAS 37149-18-1
Molecular Formula C10H18NO3
Molecular Weight (g/mol) 200.258
MDL Number MFCD00134569
InChI Key CYQGCJQJIOARKD-UHFFFAOYSA-N
Synonym 4-Carboxy-TEMPO
PubChem CID 3080786
IUPAC Name 1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine-4-carboxylic acid
SMILES CC1(CC(CC(N1[O])(C)C)C(=O)O)C

Specifications

Melting Point 178°C
Quantity 100 mg
Formula Weight 200.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Carboxy-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical