accessibility menu, dialog, popup

UserName

CAS RN 36810-87-4

SCNO

CAS RN 36810-87-4

IUPAC Name: 2-(isothiocyanatomethyl)oxolane
Synonyms: 2-(isothiocyanatomethyl)oxolane
Molecular Weight (g/mol): 143.2
Molecular Formula: C6H9NOS
InChi Key: CSFIFTGMGITVRE-UHFFFAOYNA-N
SMILES: S=C=NCC1CCCO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)