missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl isothiocyanate, 98%

Catalog No. p-7045227
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0987803 1 g
AAL0987806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0987803 Supplier Thermo Scientific Chemicals Supplier No. L0987803
Only null left

CAS: 36810-87-4 | C6H9NOS

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36810-87-4
Molecular Formula C6H9NOS
MDL Number MFCD00022476
Synonym 2-(Isothiocyanatomethyl)tetrahydrofuran

Specifications

Density 1.12
Boiling Point 122°C to 125°C (18 mmHg)
Refractive Index 1.535
Quantity 1 g
UN Number UN2922
Beilstein 111696
Sensitivity Moisture sensitive
Formula Weight 143.21
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl isothiocyanate
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only